missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16971-1-AP-150UL
This item is not returnable.
View return policy
Description
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile "growth cones" that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.Specifications
| GAP43 | |
| Polyclonal | |
| Unconjugated | |
| GAP43 | |
| Axonal membrane protein GAP 43, B 50, GAP43, growth associated protein 43, Neural phosphoprotein B 50, Neuromodulin, PP46 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14432, 2596, 29423 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06837, P07936, P17677 | |
| Gap43 | |
| GAP43 Fusion Protein Ag9294 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction