missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAP43 Monoclonal Antibody (K29024_1C6), Invitrogen™

Product Code. 30202612
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30202612 50 μL 50µg
30213100 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30202612 Supplier Invitrogen™ Supplier No. MA555647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VEKNDDDQKI EQDGIKPEDK AHKAATKIQA SFRGHITRKK LKGEKKDDVQ AAEAEANKKD EAPVADGVEK KGEGTTTAEA APATGSKPDE PGKAGETPSE

GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAP43
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K29024_1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene GAP43
Gene Accession No. P07936, P17677
Gene Alias Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Gene Symbols GAP43
Host Species Mouse
Immunogen Recombinant human GAP43 (immunogen range is 11-110).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2596, 29423
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.