missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAP43 Monoclonal Antibody (7B10)

Description
This antibody can recognize both the phosphorylated and unphosphorylated target.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
Specifications
Specifications
| Antigen | GAP43 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 7B10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | GAP43 |
| Gene Accession No. | P06837, P07936, P17677, P35001 |
| Gene Alias | Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?