Learn More
GAP-43 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 75 publications
Brand: Novus Biologicals NB300-143
This item is not returnable.
View return policy
Description
GAP-43 Polyclonal specifically detects GAP-43 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Equine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Flow Cytometry, Immunohistochemistry | |
| GAP43 | |
| RUO | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 1:10000, Flow Cytometry, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen 1-2 ug/ml | |
| 0.1 mL | |
| Pig | |
| IgG |