missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Antibody (CL10847), Novus Biologicals™
Mouse Monoclonal Antibody
£317.00 - £473.00
Specifications
| Antigen | GAP-43 |
|---|---|
| Clone | CL10847 |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602585
|
Novus Biologicals
NBP3-21181-25ul |
25 μg |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653734
|
Novus Biologicals
NBP3-21181-100ul |
100 μg |
£473.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| GAP-43 | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| This antibody was developed against a Synthetic Peptide of P17677. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL10847 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| PBS, pH 7.2, 40% glycerol | |
| 2596 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title