missing translation for 'onlineSavingsMsg'
Learn More

GAP-43 Antibody (5E8), Novus Biologicals™

Mouse Monoclonal Antibody

£146.00 - £403.00

Specifications

Antigen GAP-43
Clone 5E8
Concentration 1 mg/mL
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000
Applications Western Blot, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18672009
View Documents
Novus Biologicals
NBP2-50052SS
0.025 mL
£146.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18632709
View Documents
Novus Biologicals
NBP2-50052
0.1 mL
£403.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

GAP-43
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience
50% PBS, 50% glycerol
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Recombinant full length human GAP-43. [UniProt# P17677]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
5E8
Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000
Monoclonal
Purified
RUO
Human, Mouse, Rat
P17677
2596
IgG1
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.