missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Antibody (3H14), Novus Biologicals™
Mouse Monoclonal Antibody
£270.00 - £392.00
Specifications
| Antigen | GAP-43 |
|---|---|
| Clone | 3H14 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:1000-5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669747
|
Novus Biologicals
NBP3-05535-100ul |
100 μg |
£392.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624716
|
Novus Biologicals
NBP3-05535-50ul |
50 μg |
£270.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| GAP-43 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 50% PBS, 50% glycerol | |
| 2596 | |
| IgM | |
| Immunogen affinity purified |
| 3H14 | |
| Western Blot 1:5000, Immunohistochemistry 1:1000-5000, Immunocytochemistry/ Immunofluorescence 1:1000-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| Recombinant full-length Human GAP-43 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title