missing translation for 'onlineSavingsMsg'
Learn More

GANT 58, MedChemExpress

Product Code. 16455618
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455618

Brand: Medchem Express HY13282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GANT 58 (NSC 75503) is a potent GLI antagonist that inhibits GLI1-induced transcription with IC50 of 5 μM.

GANT 58 (NSC 75503) is a potent GLI antagonist that inhibits GLI1-induced transcription with IC50 of 5 μM.

Specifications

Chemical Name or Material GANT 58
Physical Form Solid
CAS 64048-12-0
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H16N4S
For Use With (Application) Cancer-programmed cell death
Synonym NSC 75503
Solubility Information Ethanol : 20 mg/mL (50.96 mM; Need ultrasonic) ∣DMSO : 9.09 mg/mL (23.16 mM; Need ultrasonic)
SMILES C1(C2=C(C(C3=CC=NC=C3)=C(S2)C4=CC=NC=C4)C5=CC=NC=C5)=CC=NC=C1
Molecular Weight (g/mol) 392.48
Formula Weight 392.48
Percent Purity 99.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.