missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GANP Monoclonal Antibody (5B2C4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548659
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GANP Monoclonal Antibody for Western Blot, Flow, ELISA
POLE3 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
Spécification
| GANP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O60318 | |
| MCM3AP | |
| Purified recombinant fragment of human MCM3AP (AA: 293-442) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 5B2C4 | |
| Unconjugated | |
| MCM3AP | |
| 80 kDa MCM3-associated protein; FLJ44336; FLJ45306; GANP; GC-associated DNA primase; germinal center-associated nuclear protein; germinal-center associated nuclear protein; germinal-centre associated nuclear protein; KIAA0572; MAP80; MCM3 acetylating protein; MCM3 acetyltransferase; MCM3 import protein; MCM3 minichromosome maintenance deficient 3 associated protein; MCM3AP; minichromosome maintenance complex component 3 associated protein; minichromosome maintenance deficient 3 (S. cerevisiae) associated protein; minichromosome maintenance deficient 3 associated protein; minichromosome maintenance deficient 3-associated protein; mKIAA0572; nuclear protein GANP; SAC3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu