missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gankyrin Antibody (OTI3F6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71267
This item is not returnable.
View return policy
Description
Gankyrin Monoclonal specifically detects Gankyrin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Gankyrin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Flow Cytometry 1:100, CyTOF-ready | |
| 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
| Mouse | |
| 24.4 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI3F6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PSMD10 | |
| Full length human recombinant protein of human PSMD10 (NP_002805) produced in HEK293T cell. | |
| Affinity Purified | |
| Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 5716 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction