missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gankyrin Antibody (1F4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59400FR
This item is not returnable.
View return policy
Description
Gankyrin Monoclonal antibody specifically detects Gankyrin in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Gankyrin | |
| Monoclonal | |
| DyLight 680 | |
| 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 | |
| Recombinant human Gankyrin (1-226aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway | |
| 5716 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1F4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction