missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ganirelix Trifluoroacetic Acid Salt (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Ganirelix Trifluoroacetic Acid Salt |
| Molecular Formula | C80H113ClN18O13 • x(C2HF3O2) |
| Quantity | 5 mg |
| Synonym | N-Acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-L-tyrosyl-N6-[bis(ethylamino)methylene]-D-lysyl-L-leucyl-N6-[bis(ethylamino)methylene]-L-lysyl-L-prolyl-D-alaninamide Trifluoroacetic Acid Salt, Antagon, Orgalutran Trifluoroacetic Acid Salt, RS 26306 Trifluoroacetic Acid Salt |
| SMILES | O=C(C)N[C@@H](C(N[C@H](CC1=CC=C(Cl)C=C1)C(N[C@@H](C(N[C@H](C(N[C@H](C(N[C@@H](C(N[C@H](C(N[C@@H](CCCC/N=C(NCC)\NCC)C(N2CCC[C@H]2C(N[C@H](C)C(N)=O)=O)=O)=O)CC(C)C)=O)CCCC/N=C(NCC)/NCC)=O)CC3=CC=C(O)C=C3)=O)CO)=O)CC4=CN=CC=C4)=O)=O)CC5=CC6=C(C=C5)C=CC=C6.OC(C(F)(F)F)=O.[F\,Cl\,Br\,I] |
| Molecular Weight (g/mol) | 1570.3211402 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?