missing translation for 'onlineSavingsMsg'
Learn More

Ganglioside GT1b trisodium salt, TRC

Product Code. 30394965
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394965 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394965 Supplier Toronto Research Chemicals Supplier No. TRCG2350781MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ganglioside GT1b trisodium salt
CAS 59247-13-1
Molecular Formula C95H162N5Na3O47
Quantity 1 mg
InChI Formula InChI=1S/C95H165N5O47.3Na/c1-7-9-11-13-15-17-19-21-22-24-26-28-30-32-34-36-64(118)100-52(53(112)35-33-31-29-27-25-23-20-18-16-14-12-10-8-2)47-134-87-75(125)74(124)78(62(45-106)137-87)139-89-77(127)85(147-95(92(132)133)39-56(115)67(98-50(5)110)83(145-95)73(123)61(44-105)142-93(90(128)129)37-54(113)65(96-48(3)108)81(143-93)69(119)57(116)40-101)79(63(46-107)138-89)140-86-68(99-51(6)111)80(71(121)59(42-103)135-86)141-88-76(126)84(72(122)60(43-104)136-88)146-94(91(130)131)38-55(114)66(97-49(4)109)82(144-94)70(120)58(117)41-102;;;/h33,35,52-63,65-89,101-107,112-117,119-127H,7-32,34,36-47H2,1-6H3,(H,96,108)(H,97,109)(H,98,110)(H,99,111)(H,100,118)(H,128,129)(H,130,131)(H,132,133);;;/q;3*+1/p-3/b35-33+;;;
SMILES CCCCCCCCCCCCCCCCCC(=O)NC(COC1OC(CO)C(OC2OC(CO)C(OC3OC(CO)C(O)C(OC4OC(CO)C(O)C(OC5(CC(O)C(\N=C(\C)/O[Na])C(O5)C(O)C(O)CO)C(=O)O)C4O)C3NC(=O)C)C(OC6(CC(O)C(\N=C(\C)/O[Na])C(O6)C(O)C(CO)OC7(CC(O)C(\N=C(\C)/O[Na])C(O7)C(O)C(O)CO)C(=O)O)C(=O)O)C2O)C(O)C1O)C(O)\C=C\CCCCCCCCCCCCC
IUPAC Name [(Z)-N-[6-[2-[3-acetamido-2-[4-[2-carboxy-6-[2-[2-carboxy-4-hydroxy-5-[(Z)-1-sodiooxyethylideneamino]-6-(1,2,3-trihydroxypropyl)oxan-2-yl]oxy-1,3-dihydroxypropyl]-4-hydroxy-5-[(Z)-1-sodiooxyethylideneamino]oxan-2-yl]oxy-6-[4,5-dihydroxy-2-(hydroxymethyl)-6-[(E)-3-hydroxy-2-(octadecanoylamino)octadec-4-enoxy]oxan-3-yl]oxy-5-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-6-carboxy-4-hydroxy-2-(1,2,3-trihydroxypropyl)oxan-3-yl]-C-methylcarbonimidoyl]oxysodium
Molecular Weight (g/mol) 2195.28
Formula Weight 2194.0133
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.