missing translation for 'onlineSavingsMsg'
Learn More

Ganglioside GM1 Sodium Salt (bovine brain), TRC

Product Code. 30399665
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399665 25 mg
30389502 5 mg
30391946 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399665 Supplier Toronto Research Chemicals Supplier No. TRCG23506825MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37758-47-7
Molecular Formula C73 H131 N3 O31 . x Na
Molecular Weight (g/mol) 1546.82
Synonym Ganglioside G4 Sodium Salt, GM1 Sodium Salt, GM1-Ganglioside Sodium Salt, Ganglioside A2 Sodium Salt, Ganglioside GGtet1 Sodium Salt, Ganglioside GI Sodium Salt, Ganglioside GM1a Sodium Salt, Ganglioside M1 Sodium Salt, Sialosylganglio-N-tetraosylceramide Sodium Salt, Sygen Sodium Salt
IUPAC Name sodium;(2S,4S,5R,6R)-5-acetamido-2-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5R,6R)-3-acetamido-5-hydroxy-6-(hydroxymethyl)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-tetrahydropyran-2-yl]oxy-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(E,2S,3R)-3-hydroxy-2-(octadecanoylamino)octadec-4-enoxy]tetrahydropyran-3-yl]oxy-3-hydroxy-6-(hydroxymethyl)tetrahydropyran-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]tetrahydropyran-2-carboxylic acid
SMILES CCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H]3NC(=O)C)[C@H](O[C@@]5(C[C@H](O)[C@@H](NC(=O)C)[C@@H](O5)[C@@H](O)[C@@H](O)CO)C(=O)O)[C@H]2O)[C@H](O)[C@H]1O)[C@H](O)\C=C\CCCCCCCCCCCCC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C73H131N3O31/c1-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-52(87)76-44(45(84)33-31-29-27-25-23-21-18-16-14-12-10-8-6-2)41-98-69-61(94)59(92)63(50(39-80)101-69)103-71-62(95)67(107-73(72(96)97)35-46(85)53(74-42(3)82)66(106-73)55(88)47(86)36-77)64(51(40-81)102-71)104-68-54(75-43(4)83)65(57(90)49(38-79)99-68)105-70-60(93)58(91)56(89)48(37-78)100-70/h31,33,44-51,53-71,77-81,84-86,88-95H,5-30,32,34-41H2,1-4H3,(H,74,82)(H,75,83)(H,76,87)(H,96,97)/b33-31+/t44-,45+,46-,47-,48+,49+,50+,51+,53+,54+,55-,56-,57-,58-,59+,60+,61+,62+,63+,64-,65+,66+,67+,68-,69+,70-,71-,73-/m0/s1
Formula Weight 1545.8767
Chemical Name or Material Ganglioside GM1 sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.