missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Ganglioside GD2 Antibody (EMD 273063) - Humanized, Novus Biologicals™

Product Code. 30045308 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30045308 1 mg 1mg
30045495 100 μg 100µg
30046778 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30045308 Supplier Novus Biologicals™ Supplier No. NBP3282901mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Ganglioside GD2 Monoclonal antibody specifically detects Ganglioside GD2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ganglioside GD2
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone EMD 273063
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Host Species Human
Immunogen GD2
Purification Method Protein A purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.