missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ganglioside GD2 Antibody (14G2a) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43551
This item is not returnable.
View return policy
Description
Ganglioside GD2 Monoclonal antibody specifically detects Ganglioside GD2 in Human samples. It is validated for Flow Cytometry,ELISA
Specifications
| Ganglioside GD2 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 14G2a | |
| Unconjugated | |
| PBS | |
| The original murine parent antibody was generated against human neuroblastoma cell line LAN-1. This IgG2a isotype version was then generated by class switching of the murine parent antibody 14.18 (IgG3). | |
| 0.2 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction