missing translation for 'onlineSavingsMsg'
Learn More

Ganetespib, MedChemExpress

Product Code. 16496666
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496666 100 mg 100mg
16492477 50 mg 50mg
16491166 200 mg 200mg
16456368 10 mg 10mg
16438957 5 mg 5mg
16411766 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496666 Supplier Medchem Express Supplier No. HY15205
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ganetespib (STA-9090) is a heat shock protein 90 (HSP90) inhibitor which exhibits potent cytotoxicity in a wide variety of hematological and solid tumor cell lines. Ganetespib has antiangiogenic effects in colorectal cancer mediated through inhibition of HIF-1α and STAT3.

Ganetespib (STA-9090) is a heat shock protein 90 (HSP90) inhibitor which exhibits potent cytotoxicity in a wide variety of hematological and solid tumor cell lines. Ganetespib has antiangiogenic effects in colorectal cancer mediated through inhibition of HIF-1α and STAT3.

Specifications

Chemical Name or Material Ganetespib
CAS 888216-25-9
Color White
Purity Grade Notes Research
Molecular Formula C20H20N4O3
Quantity 100 mg
Synonym STA-9090
Solubility Information DMSO : ≥ 32 mg/mL (87.82 mM)
SMILES CN1C2=CC=C(N3C(NN=C3C4=CC(C(C)C)=C(O)C=C4O)=O)C=C2C=C1
Molecular Weight (g/mol) 364.4
Formula Weight 364.4
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.