missing translation for 'onlineSavingsMsg'
Learn More

Ganciclovir Mono-O-acetate, TRC

Product Code. 30395949
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395949 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395949 Supplier Toronto Research Chemicals Supplier No. TRCG235010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ganciclovir Mono-O-acetate
CAS 88110-89-8
Molecular Formula C11 H15 N5 O5
Quantity 250 mg
Synonym 6H-Purin-6-one, 9-[[2-(acetyloxy)-1-(hydroxymethyl)ethoxy]methyl]-2-amino-1,9-dihydro-, 2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-acetoxy-1-propanol, 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]-3-hydroxypropyl acetate, Ganciclovir Mono-O-acetate, Monoacetoxyganciclovir, O-Acetoxyganciclovir, Valganciclovir Hydrochloride Imp. E (EP)
InChI Formula InChI=1S/C11H15N5O5/c1-6(18)20-3-7(2-17)21-5-16-4-13-8-9(16)14-11(12)15-10(8)19/h4,7,17H,2-3,5H2,1H3,(H3,12,14,15,19)
SMILES CC(=O)OCC(CO)OCn1cnc2C(=O)NC(=Nc12)N
IUPAC Name [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-hydroxypropyl] acetate
Molecular Weight (g/mol) 297.27
Formula Weight 297.1073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.