missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAMT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E7B11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GAMT |
|---|---|
| Clone | 1E7B11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898085
|
Proteintech
66322-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888085
|
Proteintech
66322-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAMT is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in its gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals.The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene.Specifications
| GAMT | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GAMT, PIG2, TP53I2 | |
| GAMT | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1E7B11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Pig, Human, Mouse, Rat | |
| O35969, P10868, Q14353 | |
| 100625146, 14431, 25257, 2593 | |
| GAMT Fusion Protein Ag16778 | |
| Primary | |
| -20°C | |
| GAMT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title