missing translation for 'onlineSavingsMsg'
Learn More

gamma-Undecanolactone, 99%

Product Code. 11352867
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352867 100 mL 100mL
11362867 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352867 Supplier Thermo Scientific Alfa Aesar Supplier No. L16028.AE
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104-67-6 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-67-6
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00005405
InChI Key PHXATPHONSXBIL-UHFFFAOYSA-N
Synonym gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone
PubChem CID 7714
IUPAC Name 5-heptyloxolan-2-one
SMILES CCCCCCCC1CCC(=O)O1

Specifications

Density 0.949
Boiling Point 164°C to 166°C (13 mmHg)
Flash Point 156°C (312°F)
Refractive Index 1.45
Quantity 100 mL
Beilstein 81943
Formula Weight 184.28
Percent Purity 99%
Chemical Name or Material gamma-Undecanolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.