missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma tubulin Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
This antibody can label the centrosome structure.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Specifications
Specifications
| Antigen | gamma tubulin |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | TUBG1 |
| Gene Accession No. | P23258, P83887, Q9GKK5 |
| Gene Alias | Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma |
| Gene Symbols | TUBG1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?