Learn More
gamma tubulin Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26195-1-AP-150UL
 View more versions of this product
Product Code. 16854715
  • £349.00 / 150µL
Description
This antibody can label the centrosome structure.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Specifications
Specifications
gamma tubulin
Polyclonal
Unconjugated
TUBG1
Gamma 1 tubulin, GCP 1, TUBG, TUBG1, TUBGCP1, Tubulin gamma 1 chain, tubulin, gamma 1, tubulin-gamma
Rabbit
Antigen Affinity Chromatography
RUO
103733, 403695, 7283
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23258, P83887, Q9GKK5
TUBG1
gamma tubulin Fusion Protein Ag23575
150 μL
Primary
Canine, Human, Mouse