missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Synuclein Recombinant Rabbit Monoclonal Antibody (SAIC-31A-10)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547912
This item is not returnable.
View return policy
Description
Specificity: Recognises human SNCG.
This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
Specifications
| gamma Synuclein | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O76070 | |
| SNCG | |
| The KLH-conjugated peptide TVEEAENIAVTSGVVR, derived from Gamma-synuclein. Gamma-synulein plays a role in neurofilament network integrity. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| SAIC-31A-10 | |
| Unconjugated | |
| SNCG | |
| BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction