missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Synuclein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589862
This item is not returnable.
View return policy
Description
Immunogen sequence: QSVTSVAEKT KEQANAVSEA VVSSVNTVAT KTVEEAENIA VTSGVVRKED LRPSAPQQEG EASKEKEEVA EEAQSGGD; Positive Samples: SKOV3, HT-29; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, perinuclear region, spindle.
This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
Specifications
| gamma Synuclein | |
| Polyclonal | |
| Unconjugated | |
| SNCG | |
| BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20618, 6623 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O76070, Q9Z0F7 | |
| SNCG | |
| A synthetic peptide corresponding to a sequence within amino acids 50-127 of human gamma-Synuclein (NP_0030782). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction