missing translation for 'onlineSavingsMsg'
Learn More

gamma-secretase modulator 2, MedChemExpress

Product Code. 16498098
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498098 10 mg 10mg
16474017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498098 Supplier Medchem Express Supplier No. HY50754
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress gamma-secretase modulator 2 is a potent and selective (gamma)-secretase modulator for treatment of Alzheimer(acute)s disease.

gamma-secretase modulator 2 is a potent and selective (gamma)-secretase modulator for treatment of Alzheimer(acute)s disease.

Specifications

Chemical Name or Material gamma-secretase modulator 2
CAS 1093978-89-2
Color White
Purity Grade Notes Research
Molecular Formula C25H22F4N6O2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (194.37 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=CC(F)=C2CCC1N3N=NC(C4=CC(OC)=C(N5C=NC(C)=C5)C=C4)=C3)CC(F)(F)F
Molecular Weight (g/mol) 514.47
Formula Weight 514.47
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.