missing translation for 'onlineSavingsMsg'
Learn More

gamma-Polyglutamic Acid Sodium Salt (Technical Grade), TRC

Product Code. 30421022
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421022 2.5 g 2.5g
30421021 25 g -
30425441 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421022 Supplier Toronto Research Chemicals Supplier No. TRCP6894452.5G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26247-79-0
Molecular Formula (C5H8NNaO4)n
Synonym Glutamic acid homopolymer sodium salt, L-Glutamic acid polymer sodium salt
SMILES N[C@@H](CCC(O[Na])=O)C(O)=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H9NO4.Na/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);/t3-;/m0./s1
Chemical Name or Material gamma-Polyglutamic Acid Sodium Salt (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.