missing translation for 'onlineSavingsMsg'
Learn More

gamma-Polyglutamic Acid Sodium Salt (Technical Grade), TRC

Product Code. 30421021
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421021 25 g -
30421022 2.5 g 2.5g
30425441 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421021 Supplier Toronto Research Chemicals Supplier No. TRCP68944525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26247-79-0
Molecular Formula (C5H8NNaO4)n
Synonym Glutamic acid homopolymer sodium salt, L-Glutamic acid polymer sodium salt
SMILES N[C@@H](CCC(O[Na])=O)C(O)=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H9NO4.Na/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);/t3-;/m0./s1
Chemical Name or Material gamma-Polyglutamic Acid Sodium Salt (Technical Grade)
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.