missing translation for 'onlineSavingsMsg'
Learn More

Gamma-Lipoic Acid, TRC

Product Code. 30471449
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471449 500 mg 500mg
30469458 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30471449 Supplier Toronto Research Chemicals Supplier No. TRCL468755500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 462-20-4
Molecular Formula H2 O(C8 H14 O S2)n
Molecular Weight (g/mol) 208.3414
Synonym Octanoic acid, 6,8-dimercapto-, 6,8-Dimercaptooctanoic acid, (±)-Dihydro-α-lipoic acid, (±)-Dihydrolipoic acid, 6,8-Dihydrothioctic acid, 6,8-Dithiooctanoic acid, DL-Dihydro-α-lipoic acid, Dihydrothioctic acid, Reduced lipoic acid, Reduced thioctic acid, Thioctic acid, dihydro-, dl-Dihydrolipoic acid, α-Lipoic acid, dihydro-, γ-Lipoic acid, Thioctic Acid Impurity 2
IUPAC Name 6,8-bis(sulfanyl)octanoic acid
SMILES OC(=O)CCCCC(S)CCS

Tekniske data

Quantity 500 mg
InChI Formula InChI=1S/C8H16O2S2/c9-8(10)4-2-1-3-7(12)5-6-11/h7,11-12H,1-6H2,(H,9,10)
Formula Weight 208.0592
Chemical Name or Material Dihydrolipoic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.