missing translation for 'onlineSavingsMsg'
Learn More

gamma-Hexanolactone, 98%

Product Code. 11474187
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474187 100 g 100g
11464187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474187 Supplier Thermo Scientific Alfa Aesar Supplier No. A19642.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-06-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005401
InChI Key JBFHTYHTHYHCDJ-UHFFFAOYNA-N
Synonym gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one
PubChem CID 12756
ChEBI CHEBI:85235
IUPAC Name 5-ethyloxolan-2-one
SMILES CCC1CCC(=O)O1

Specifications

Melting Point -18°C
Density 1.025
Boiling Point 218°C to 219°C
Flash Point 98°C (208°F)
Refractive Index 1.439
Quantity 100 g
Beilstein 107260
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material gamma-Hexanolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.