missing translation for 'onlineSavingsMsg'
Learn More

gamma-Glutamylcyclotransferase/CRF21/GGCT Antibody (04), PerCP, Novus Biologicals™

Product Code. 18630718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18630718 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18630718 Supplier Novus Biologicals Supplier No. NBP306551PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

gamma-Glutamylcyclotransferase/CRF21/GGCT Monoclonal antibody specifically detects gamma-Glutamylcyclotransferase/CRF21/GGCT in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma-Glutamylcyclotransferase/CRF21/GGCT
Applications ELISA
Classification Monoclonal
Clone 04
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias gamma-glutamylcyclotransferase, GCTG, GGC, MGC3077
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human gamma-Glutamylcyclotransferase/CRF21/GGCT (Uniprot#: O75223-1; Met1-Leu188).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 79017
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.