missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Glutamylcyclotransferase/CRF21/GGCT Antibody (04), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | gamma-Glutamylcyclotransferase/CRF21/GGCT |
|---|---|
| Clone | 04 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611167
|
Novus Biologicals
NBP3-06551-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625916
|
Novus Biologicals
NBP3-06551-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma-Glutamylcyclotransferase/CRF21/GGCT Monoclonal antibody specifically detects gamma-Glutamylcyclotransferase/CRF21/GGCT in Human samples. It is validated for ELISASpecifications
| gamma-Glutamylcyclotransferase/CRF21/GGCT | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| gamma-glutamylcyclotransferase, GCTG, GGC, MGC3077 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human gamma-Glutamylcyclotransferase/CRF21/GGCT (Uniprot#: O75223-1; Met1-Leu188). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 04 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, Endocrinology | |
| 0.2 um filtered solution in PBS | |
| 79017 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title