missing translation for 'onlineSavingsMsg'
Learn More

gamma-glutamyl hydrolase Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18355712 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355712 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355712 Supplier Novus Biologicals Supplier No. NBP306418AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

gamma-glutamyl hydrolase Polyclonal antibody specifically detects gamma-glutamyl hydrolase in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma-glutamyl hydrolase
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Conjugase, EC 3.4.19.9, gamma-glutamyl hydrolase, gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase), Gamma-Glu-X carboxypeptidase, GH
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human gamma-glutamyl hydrolase fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8836
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.