missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma-ENaC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109307
This item is not returnable.
View return policy
Description
Positive Control: A431, 293T, rat kidney tissue, mouse brain tissue.
Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel's syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications
| gamma-ENaC | |
| Polyclonal | |
| Unconjugated | |
| SCNN1G | |
| [g]ENaC; amiloride sensitive sodium channel gamma1 subunit; amiloride-sensitive epithelial sodium channel gamma subunit; amiloride-sensitive sodium channel gamma-subunit; amiloride-sensitive sodium channel subunit gamma; BESC3; ENaC; ENaC gamma; ENaC gamma subunit; ENaCG; ENaCgamma; epithelial Na(+) channel subunit gamma; epithelial sodium channel gamma subunit; Gamma ENaC; Gamma NaCH; gamma-ENaC; Gamma-NaCH; nonvoltage-gated sodium channel 1 subunit gamma; PHA 1; PHA1; SCNEG; SCNN 1G; Scnn1g; sodium channe epithelial 1 gamma subunit; sodium channel epithelial 1 gamma subunit; Sodium channel nonvoltage-gated 1 gamma (epithelial); sodium channel, non voltage gated 1 gamma subunit; sodium channel, nonvoltage-gated 1 gamma; sodium channel, nonvoltage-gated 1, gamma; Sodium channel, nonvoltage-gated 1, gamma (epithelial); sodium channel, non-voltage-gated 1, gamma subunit; sodium channel, nonvoltage-gated, type I, gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6340 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P51170 | |
| SCNN1G | |
| Recombinant protein within Human SCNN1G aa 77-541. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction