missing translation for 'onlineSavingsMsg'
Learn More

gamma-DGG, MedChemExpress

Product Code. p-200099955
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475897

Brand: Medchem Express HY100785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress gamma-DGG is a competitive AMPA receptor blocker.

gamma-DGG is a competitive AMPA receptor blocker.

Specifications

Chemical Name or Material gamma-DGG
Physical Form Solid
CAS 6729-55-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C7H12N2O5
For Use With (Application) Neuroscience-Neuromodulation
Synonym γDGG γ-D-Glutamylglycine
Solubility Information H2O : ≥ 150 mg/mL (734.65 mM)
SMILES OC(CNC(CC[C@@H](N)C(O)=O)=O)=O
Molecular Weight (g/mol) 204.18
Formula Weight 204.18
Percent Purity 97.17%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.