missing translation for 'onlineSavingsMsg'
Learn More

gamma-Decanolactone, 98%

Product Code. 11468900
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468900 5 g 5g
11448617 25 g 25g
11458617 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468900 Supplier Thermo Scientific Alfa Aesar Supplier No. A19732.06
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 706-14-9 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706-14-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00005404
InChI Key IFYYFLINQYPWGJ-UHFFFAOYNA-N
Synonym gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
PubChem CID 12813
IUPAC Name 5-hexyloxolan-2-one
SMILES CCCCCCC1CCC(=O)O1

Specifications

Density 0.948
Boiling Point 150°C to 153°C (15 mmHg)
Flash Point 146°C (294°F)
Refractive Index 1.5
Quantity 5 g
Beilstein 117547
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material gamma-Decanolactone
compliance-icons
Product Identifier
  • gamma-Decalactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: gamma-Decalactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.