missing translation for 'onlineSavingsMsg'
Learn More

Gamma-D-Glutamylaminomethylsulfonic Acid, TRC

Product Code. 30400543
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400543 25 mg
30408043 50 mg
30390883 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400543 Supplier Toronto Research Chemicals Supplier No. TRCG59729325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90237-02-8
Molecular Formula C6H12N2O6S
Molecular Weight (g/mol) 240.23
Synonym N-(Sulfomethyl)-D-glutamine, GAMS, gamma-D-Glutamylaminomethyl sulfonate
IUPAC Name (2R)-2-amino-5-oxo-5-(sulfomethylamino)pentanoic acid
SMILES N[C@H](CCC(=O)NCS(=O)(=O)O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H12N2O6S/c7-4(6(10)11)1-2-5(9)8-3-15(12,13)14/h4H,1-3,7H2,(H,8,9)(H,10,11)(H,12,13,14)/t4-/m1/s1
Formula Weight 240.0416
Chemical Name or Material gamma-D-Glutamylaminomethylsulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.