missing translation for 'onlineSavingsMsg'
Learn More

gamma catenin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen gamma catenin
Concentration 0.2 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16825885
View Documents
Proteintech
27872-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16815885
View Documents
Proteintech
27872-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

JUP is a major cytoplasmic protein which is the only known constituent common to submembranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
Specifications

Specifications

gamma catenin
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Unconjugated
Rabbit
Human
P14923
3728
gamma catenin Fusion Protein Ag27394
Primary
-20°C
JUP
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ARVD12, Catenin gamma, CTNNG, Desmoplakin 3, Desmoplakin III, DP3, DPIII, junction plakoglobin, JUP, PDGB, PKGB, gamma-Catenin
JUP
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.