Learn More
Invitrogen™ gamma Catenin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595204
Description
Human gamma Catenin shares 98% amino acid (aa) sequence identity with both mouse and rat gamma Catenin. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human MCF-7 whole cell, human T47D whole cell, rat heart tissue. IHC: human intestinal cancer tissue, mouse intestine tissue, rat intestine tissue IHC-F: rat intestine tissue. ICC/IF: A431 cell, human MCF-7 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
Specifications
| gamma Catenin | |
| Polyclonal | |
| Unconjugated | |
| JUP | |
| ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16480, 3728, 81679 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P14923, Q02257, Q6P0K8 | |
| JUP | |
| E.coli-derived human gamma Catenin recombinant protein (Position: M556-A745). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.