missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Catenin Monoclonal Antibody (PG-11E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 138500
This item is not returnable.
View return policy
Description
gamma Catenin Monoclonal Antibody for ICC/IF, IHC, IP, Western Blot
JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
Specifications
| gamma Catenin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P14923, Q02257 | |
| JUP | |
| Fusion protein consisting of the maltose binding protein fused to full length human plakoglobin protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| PG-11E4 | |
| Unconjugated | |
| JUP | |
| ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 16480, 3728 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction