missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Antibody (CTNG/1664), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £409.00
Specifications
| Antigen | gamma Catenin |
|---|---|
| Clone | CTNG/1664 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034936
|
Novus Biologicals
NBP2-53172-20UG |
20 μg |
£222.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257940
|
Novus Biologicals
NBP2-53172-100UG |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| gamma Catenin | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers, Phospho Specific | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 3728 | |
| Recombinant human full-length gamma catenin protein | |
| Primary | |
| Store at 4C. |
| CTNG/1664 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| JUP | |
| IgG1 κ | |
| Protein A or G purified | |
| 84 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title