missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Catenin Antibody (CTNG/1483), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54525AF647
This item is not returnable.
View return policy
Description
gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| gamma Catenin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| Recombinant human gamma Catenin protein (Uniprot: P14923) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 3728 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CTNG/1483 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction