Learn More
Polycarbin Gamma Carbin Program for Nitrile Gloves (5-Pack)
GREENER_CHOICE
Product Code. 31702904
Change view
Click to view available options
Product Type:
Recycling and Sustainability
Unit Size:
Pack of 5
Product Code. Product Type unitSize
31702904 Recycling and Sustainability Pack of 5
1 options
Product Code. 31702904 Supplier Polycarbin Supplier No. PR010301UK
Edge
Closed-loop recycling for non-hazardous nitrile gloves, helping labs achieve verified sustainability and waste reduction.
  • Closed-Loop Recycling for Nitrile Gloves: Provides laboratories with a simple recycling program for non-hazardous, non-infectious nitrile gloves, diverting waste from landfill and incineration while promoting circular resource use.
  • Easy Mailback Process: Each kit includes five (5) 61-liter (16-gallon) collection containers with prepaid return labels, making glove collection and return seamless across laboratory operations.
  • Universal Compatibility: Accepts nitrile gloves from all manufacturers, allowing flexible participation regardless of supplier or glove brand.
  • Safe and Compliant: Designed exclusively for gloves free from hazardous or biological contamination, ensuring safe handling and adherence to waste transport regulations.
  • Verified Impact Tracking: Includes access to Polycarbin's Carbin Counter™ platform, which delivers ISO 14040/14044-compliant, third-party verified data on waste diversion, carbon reduction, and circular recovery metrics.
  • Supports ESG and Scope 3 Goals: Recycled nitrile materials are transformed into high-quality, low-carbon products that help institutions meet sustainability and procurement objectives.
  • Cost and Carbon Savings: Reduces waste disposal costs while minimizing greenhouse gas emissions associated with traditional waste management.
  • Laboratory Integration: Fits seamlessly into existing waste segregation workflows and complies with regulated laboratory standards for safe operation.
  • Convenient Configuration: Multi-container design supports efficient distribution across lab areas for easy collection and participation.
  • Proven Circular Outcome: Ensures that recovered nitrile materials are reprocessed into durable, sustainable products, closing the loop on glove waste.
Specifications
Product Type Recycling and Sustainability