missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
gamma C Crystallin Monoclonal antibody specifically detects gamma C Crystallin in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | gamma C Crystallin |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 |
| Formulation | PBS (pH 7.3) 1% BSA, 50% glycerol |
| Gene Alias | CCL, CRYG3gamma-C-crystallin, crystallin, gamma C, crystallin, gamma-3, Gamma-C-crystallin, Gamma-crystallin 2-1, Gamma-crystallin 3, gamma-crystallin C |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human CRYGC (NP_066269) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?