missing translation for 'onlineSavingsMsg'
Learn More

gamma C Crystallin Antibody (OTI1E3) [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30688389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688389 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688389 Supplier Novus Biologicals Supplier No. NBP344855AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

gamma C Crystallin Monoclonal antibody specifically detects gamma C Crystallin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma C Crystallin
Applications Western Blot
Classification Monoclonal
Clone OTI1E3
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CCL, CRYG3gamma-C-crystallin, crystallin, gamma C, crystallin, gamma-3, Gamma-C-crystallin, Gamma-crystallin 2-1, Gamma-crystallin 3, gamma-crystallin C
Host Species Mouse
Immunogen Full length human recombinant protein of human CRYGC (NP_066269) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 1420
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.