missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma C Crystallin Antibody (OTI1D6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | gamma C Crystallin |
|---|---|
| Clone | OTI1D6 |
| Dilution | Western Blot 1:4000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306717
|
Novus Biologicals
NBP3-44856-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306523
|
Novus Biologicals
NBP3-44856-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma C Crystallin Monoclonal antibody specifically detects gamma C Crystallin in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| gamma C Crystallin | |
| Western Blot 1:4000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CCL, CRYG3gamma-C-crystallin, crystallin, gamma C, crystallin, gamma-3, Gamma-C-crystallin, Gamma-crystallin 2-1, Gamma-crystallin 3, gamma-crystallin C | |
| Full length human recombinant protein of human CRYGC(NP_066269) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI1D6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Vision | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 1420 | |
| IgG2a | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title