Learn More
gamma-Butyrolactone, 99+, Thermo Scientific™
Chemical Identifiers
| CAS | 96-48-0 |
|---|---|
| Molecular Formula | C4;H6;O2; |
| Molecular Weight (g/mol) | 86.09 |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 96-48-0 | |
| C4;H6;O2; | |
| 86.09 |
Specifications
| 96-48-0 | |
| 204.0°C to 205.0°C | |
| 99.0 % min. (GC) | |
| C4;H6;O2; | |
| 17, 234 | |
| 1.1200 | |
| 86.09 | |
| γ-Butyrolactone |
| 1.1200 g/mL | |
| 98°C | |
| Glass bottle | |
| 1.4355 to 1.4375 | |
| 15, 1600 | |
| Solubility aq. soln.: miscible. Other solubilities: soluble in acetone, benzene, ethyl ether, ethanol,, methanol | |
| 86.09 |
Safety and Handling
GHS H Statement Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-509-5
- y-Butyrolactone
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.