missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamma adducin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17585-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Adducins are heteromeric proteins composed of different subunits referred to as adducin alpha, beta and gamma. The three subunits are encoded by distinct genes and belong to a family of membrane skeletal proteins involved in the assembly of spectrin-actin network in erythrocytes and at sites of cell-cell contact in epithelial tissues. While adducins alpha and gamma are ubiquitously expressed, the expression of adducin beta is restricted to brain and hematopoietic tissues. Adducin, originally purified from human erythrocytes, was found to be a heterodimer of adducins alpha and beta. Polymorphisms resulting in amino acid substitutions in these two subunits have been associated with the regulation of blood pressure in an animal model of hypertension. Heterodimers consisting of alpha and gamma subunits have also been described. Structurally, each subunit is comprised of two distinct domains. The amino-terminal region is protease resistant and globular in shape, while the carboxy-terminal region is protease sensitive. The latter contains multiple phosphorylation sites for protein kinase C, the binding site for calmodulin, and is required for association with spectrin and actin. Alternatively spliced adducin gamma transcripts encoding different isoforms have been described. The functions of the different isoforms are not known.Specifications
Gamma adducin | |
Polyclonal | |
Unconjugated | |
ADD3 | |
ADD3, ADDL, adducin 3 (gamma), Adducin like protein 70, Gamma adducin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
120, 25230, 27360 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62847, Q9QYB5, Q9UEY8 | |
Add3 | |
Gamma adducin Fusion Protein Ag11318 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |