missing translation for 'onlineSavingsMsg'
Learn More

Gamma Adaptin Rabbit anti-Human, Mouse, Rat, Clone: 4U2C8, Novus Biologicals™

Product Code. 18348603 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348603 100 μg 100µL
18396225 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18348603 Leverantör Novus Biologicals Leverantörsnummer NBP315727100UL

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Monoclonal Antibody

Gamma Adaptin Monoclonal antibody specifically detects Gamma Adaptin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Gamma Adaptin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4U2C8
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Adapter-related protein complex 1 subunit gamma-1, Adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTGclathrin assembly protein complex 1 gamma large chain, AP-1 complex subunit gamma-1, CLAPG1clathrin-associated/assembly/adaptor protein, large, gamma 1, Clathrin assembly protein complex 1 gamma-1 large chain, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, Golgi adaptor HA1/AP1 adaptin subunit gamma-1, MGC18255
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 723-822 of human Gamma Adaptin (O43747). RSNTNPSVTVITIQASNSTELDMTDFVFQAAVPKTFQLQLLSPSSSIVPAFNTGTITQVIKVLNPQKQQLRMRIKLTYNHKGSAMQDLAEVNNFPPQSWQ
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 164
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.