missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gamma Adaptin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144984
This item is not returnable.
View return policy
Description
Immunogen sequence: VPELMEMFLP ATKNLLNEKN HGVLHTSVVL LTEMCERSPD MLAHFRKNEK LVPQLVRILK NLIMSGYSPE HDVSGI Antigen sequence identity: Mouse 100%; Rat 51%.
Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| gamma Adaptin | |
| Polyclonal | |
| Unconjugated | |
| AP1G1 | |
| AA409002; adapter-related protein complex 1 subunit gamma-1; adaptor protein complex AP-1 gamma 1 subunit; adaptor protein complex AP-1 subunit gamma-1; adaptor protein complex AP-1, gamma 1 subunit; adaptor related protein complex 1 gamma 1 subunit; adaptor related protein complex 1 subunit gamma 1; adaptor-related protein complex 1 gamma 1 subunit; adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; ADTG; AP-1 complex subunit gamma-1; Ap1g1; AU041323; AW551707; CLAPG1; clathrin assembly protein complex 1 gamma large chain; clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; D8Ertd374e; gamma adaptin; Gamma1-adaptin; gamma-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; golgi adaptor HA1/AP1 adaptin subunit gamma-1; MGC18255; testicular tissue protein Li 21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43747 | |
| AP1G1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 168-240. Recombinant protein control fragment (Product #RP-110193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction