missing translation for 'onlineSavingsMsg'
Learn More

Gamitrinib TPP hexafluorophosphate, MedChemExpress

Product Code. 16450969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16450969

Brand: Medchem Express HY102007A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gamitrinib TPP hexafluorophosphate is a Gamitrinib (GA) mitochondrial matrix inhibitor. Gamitrinib TPP hexafluorophosphate is a mitochondrial targeted HSP90 inhibitor with anti-cancer activity.

Gamitrinib TPP hexafluorophosphate is a Gamitrinib (GA) mitochondrial matrix inhibitor. Gamitrinib TPP hexafluorophosphate is a mitochondrial targeted HSP90 inhibitor with anti-cancer activity.

Specifications

Chemical Name or Material Gamitrinib TPP hexafluorophosphate
Physical Form Solid
CAS 1131626-47-5
Color Pale Purple
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C52H65F6N3O8P2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (48.26 mM; Need ultrasonic)
SMILES F[P-](F)(F)(F)(F)F.O=C(C=C1NC(/C(C)=C/C=C/[C@H](OC)[C@@H](OC(N)=O)/C(C)=C/[C@H](C)[C@H]2O)=O)C(NCCCCCC[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)=C(C[C@H](C[C@@H]2OC)C)C1=O
Molecular Weight (g/mol) 1036.03
Formula Weight 1036.03
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.